Learn More
RAB2A Mouse anti-Human, Unconjugated, Clone: 3E1B10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67501-1-IG-150UL
Description
The RAB2A antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RAB2A. This antibody recognizes human, mouse, rat antigen. The RAB2A antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| RAB2A | |
| Monoclonal | |
| 500 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Western Blot, Immunohistochemistry, ELISA | |
| 3E1B10 | |
| Unconjugated | |
| 150 μL | |
| 5862 | |
| IgG2b |