Learn More
RAB2A Mouse anti-Human, Unconjugated, Clone: 3E1B10, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67501-1-IG-20UL
 View more versions of this product
Product Code. 16860166
  • 145.00€ / 20µL
Description
The RAB2A antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RAB2A. This antibody recognizes human, mouse, rat antigen. The RAB2A antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
RAB2A
Monoclonal
500 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunohistochemistry, ELISA
3E1B10
Unconjugated
20 μL
5862
IgG2b