Learn More
RAB37 Monoclonal Antibody (OTI2F8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA505338
Encompass_Preferred
 View more versions of this product
Product Code. 17513672
  • 540.87€ / 100µL
Description
Rab proteins are low molecular mass GTPases that are critical regulators of vesicle trafficking. For additional background information on Rab proteins.
Specifications
Specifications
RAB37
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q96AX2
RAB37
Full length human recombit protein of human RAB37 produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG2a
Immunocytochemistry, Immunofluorescence, Western Blot
OTI2F8
Unconjugated
RAB37
RAB37, member RAS oncogene family
Mouse
Affinity Chromatography
RUO
326624
-20° C, Avoid Freeze/Thaw Cycles