Learn More
RAB37 Monoclonal Antibody (OTI3B2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA505309
Description
Rab proteins are low molecular mass GTPases that are critical regulators of vesicle trafficking. For additional background information on Rab proteins.Specifications
| RAB37 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q96AX2 | |
| RAB37 | |
| Full length human recombit protein of human RAB37 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI3B2 | |
| Unconjugated | |
| RAB37 | |
| RAB37, member RAS oncogene family | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 326624 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.