missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rab3D Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
187.00€
Specifications
| Antigen | Rab3D |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 2.0 μg/mL, Immunocytochemistry/Immunofluorescence 1:10-1:500 |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
Rab3D Polyclonal specifically detects Rab3D in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| Rab3D | |
| Western Blot 2.0 μg/mL, Immunocytochemistry/Immunofluorescence 1:10-1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse | |
| D2-2, GOVras-related protein Rab-3D, RAB16glioblastoma overexpressed, RAB3D, member RAS oncogene family, RAD3DRab3D upregulated with myeloid differentiation | |
| RAB3D | |
| IgG | |
| Protein A purified | |
| Detects recombinant mouse rab 3D. This does not detect rab 3A, 3B, or 3C. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Membrane Trafficking and Chaperones, Signal Transduction | |
| PBS with 0.05% Sodium Azide | |
| 9545 | |
| Synthetic peptide corresponding to residues M(1) A S A S E P P A S P R D A A D Q N(18) of rat RAB3D. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title