missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAB5 Recombinant Rabbit Monoclonal Antibody (24GB4525)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557664
This item is not returnable.
View return policy
Description
RAB5 Recombinant Monoclonal Antibody for Western Blot, Flow
RAB5 are among the small GTPases Rab are key regulators of intracellular membrane trafficking, from the formation of transport vesicles to their fusion with membranes. Rabs cycle between an inactive GDP-bound form and an active GTP-bound form that is able to recruit to membranes different sets of downstream effectors directly responsible for vesicle formation, movement, tethering and fusion. RAB5A is required for the fusion of plasma membranes and early endosomes. Contributes to the regulation of filopodia extension. Required for the exosomal release of SDCBP, CD63, PDCD6IP and syndecan. Regulates maturation of apoptotic cell-containing phagosomes, probably downstream of DYN2 and PIK3C3.
Specifications
| RAB5 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| RAB5A | |
| 2410015H04Rik; AI663973; AU021172; nnyRab5a; Rab 5; Rab 5A; RAB5; RAB5A; RAB5A, member RAS oncogene family; Ras related protein; RAS-associated protein RAB5A; ras-related protein Rab-5A; Small GTP-binding protein rab5 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 5868 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB4525 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P20339 | |
| RAB5A | |
| A synthesized peptide derived from human Rab5 (aa 198-211). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction