missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rab7a Antibody (10E4), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-60237APC
This item is not returnable.
View return policy
Description
Rab7a Monoclonal antibody specifically detects Rab7a in Human samples. It is validated for Flow Cytometry
Specifications
| Rab7a | |
| Monoclonal | |
| APC | |
| CMT2B, FLJ20819, member RAS oncogene family, PSN, RAB7A, member RAS oncogene family, Ras-associated protein RAB7, ras-related protein Rab-7a | |
| Recombinant human RAB7A (1-207aa) purified from E. coli | |
| 0.1 mL | |
| Cellular Markers, Endosome Markers, Neuroscience, Signal Transduction | |
| 7879 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Flow Cytometry | |
| 100000 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction