Learn More
Invitrogen™ RAB7A Recombinant Rabbit Monoclonal Antibody (SN202-03)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA532340
Description
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
RAB family members are small, RAS-related GTP-binding proteins that are important regulators of vesicular transport. Each RAB protein targets multiple proteins that act in exocytic / endocytic pathways. This gene encodes a RAB family member that regulates vesicle traffic in the late endosomes and also from late endosomes to lysosomes. This encoded protein is also involved in the cellular vacuolation of the VacA cytotoxin of Helicobacter pylori. Mutations at highly conserved amino acid residues in this gene have caused some forms of Charcot-Marie-Tooth (CMT) type 2 neuropathies.
Specifications
| RAB7A | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 40% Glycerol, 0.05% BSA and 0.05% sodium azide; pH 7.4 | |
| P09527, P51149, P51150 | |
| Rab7, RAB7A | |
| Synthetic peptide within Human RAB7 aa 158-207. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot | |
| SN202-03 | |
| Unconjugated | |
| RAB7A | |
| GTP-binding protein (rab7); hypothetical protein LOC509970; PRO2706; RAB7; rab-7; RAB7, member RAS oncogene family; Rab7a; RAB7A, member RAS oncogene family; Ras-associated protein RAB7; ras-related protein BRL-RAS; Ras-related protein p23; ras-related protein Rab-7a | |
| Rabbit | |
| Protein A | |
| RUO | |
| 110315128, 19349, 29448, 7879 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.