Learn More
RAB8A Mouse anti-Human, Unconjugated, Clone: 1D12G4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67536-1-IG-150UL
Description
The RAB8A antibody from Proteintech is a mouse monoclonal antibody generated with peptide of human RAB8A. This antibody recognizes human antigen. The RAB8A antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| RAB8A | |
| Monoclonal | |
| 507 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Western Blot, ELISA | |
| 1D12G4 | |
| Unconjugated | |
| 150 μL | |
| 4218 | |
| IgG1 |