Learn More
ACVRL1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ACVRL1.
Brand: Abnova PAB19782.100ug
Description
This gene encodes a type I cell-surface receptor for the TGF-beta superfamily of ligands. It shares with other type I receptors a high degree of similarity in serine-threonine kinase subdomains, a glycine- and serine-rich region (called the GS domain) preceding the kinase domain, and a short C-terminal tail. The encoded protein, sometimes termed ALK1, shares similar domain structures with other closely related ALK or activin receptor-like kinase proteins that form a subfamily of receptor serine/threonine kinases. Mutations in this gene are associated with hemorrhagic telangiectasia type 2, also known as Rendu-Osler-Weber syndrome 2. [provided by RefSeq
Specifications
| ACVRL1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of ACVRL1. | |
| In serum (2% BSA, 0.02% sodium azide) | |
| NM_000020 | |
| ACVRL1 | |
| A synthetic peptide corresponding to C-terminus of human ACVRL1. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| ELISA (1:1000-1:10000) Western Blot (1:200-5000) Immunohistochemistry (1:100-300) The optimal working dilution should be determined by the end user. | |
| ACVRL1 | |
| ACVRLK1/ALK-1/ALK1/HHT/HHT2/ORW2/SKR3/TSR-I | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 94 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only