missing translation for 'onlineSavingsMsg'
Learn More

CYP2C8/CYP2C9/CYP2C18/CYP2C19, Rabbit, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of CYP2C8/CYP2C9/CYP2C18/CYP2C19.

Brand:  Abnova PAB24906.100uL

Product Code. 16015137

  • 503.00€ / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

CYP2C8/CYP2C9/CYP2C18/CYP2C19
Polyclonal
Unconjugated
Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:200) Immunofluorescence (1:50-1:200) The optimal working dilution should be determined by the end user.
CPC8/MP-12/MP-20
Rabbit
Antigen affinity purification
RUO
1558/1559/1562/1557
Human
Liquid
Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
1 mg/mL
Rabbit polyclonal antibody raised against synthetic peptide of CYP2C8/CYP2C9/CYP2C18/CYP2C19.
In PBS, pH 7.2 (0.05% sodium azide)
CYP2C8
A synthetic peptide corresponding to CYP2C8/CYP2C9/CYP2C18/CYP2C19.
100 μL
Primary
CYP2C8/CYP2C9/CYP2C18/CYP2C19 polyclonal antibody detects endogenous levels of CYP2C8/CYP2C9/CYP2C18/CYP2C19 protein.
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only