Learn More
FOS (phospho T232), Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of human FOS.
Brand: Abnova PAB29224.100uL
Description
The Fos gene family consists of 4 members: FOS, FOSB, FOSL1, and FOSL2. These genes encode leucine zipper proteins that can dimerize with proteins of the JUN family, thereby forming the transcription factor complex AP-1. As such, the FOS proteins have been implicated as regulators of cell proliferation, differentiation, and transformation. In some cases, expression of the FOS gene has also been associated with apoptotic cell death. [provided by RefSeq
Specifications
| FOS | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of human FOS. | |
| In PBS, pH 7.4 (without mg2+ and Ca2+), (50% glycerol, 0.02% sodium azide) | |
| P01100 | |
| FOS | |
| Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding T232 of human FOS. | |
| 100 μL | |
| Primary | |
| This antibody detects endogenous levels of FOS only when phosphorylated at threonine 232. | |
| Store at -20°C. Avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:100) Western Blot (1:500-1000) The optimal working dilution should be determined by the end user. | |
| FOS | |
| AP-1/C-FOS | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 2353 | |
| Human, Mouse | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only