missing translation for 'onlineSavingsMsg'
Learn More

Rabbit anti-Mouse IgG Fc Secondary Antibody, PE, Novus Biologicals™

Product Code. 30230910 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.5 mL
Unit Size:
0.5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30230910 0.5 mL 0.5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30230910 Supplier Novus Biologicals Supplier No. NBP260665PE
  • ONLINE EXCLUSIVE PRICE
    399.00€
    376.95€ / 0.5mL
    Save 22.05€
    5% Off
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IgG Fc Polyclonal specifically detects IgG Fc in Mouse samples. It is validated for ELISA, Immunohistochemistry, Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG Fc
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate PE
Formulation PBS
Host Species Rabbit
Immunogen Rabbit anti-Mouse IgG Fc fragment Secondary Antibody was produced by repeated immunization with mouse IgG F(c) fragment in rabbit.
Purification Method Immunogen affinity purified
Quantity 0.5 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.