Learn More
Invitrogen™ Rabbit anti-Mouse IgG (H+L) Secondary Antibody
Product Code. 15306341
Change view
Click to view available options
Quantity:
20 mg
Unit Size:
20mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15306341 20 mg 20mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15306341 Supplier Invitrogen™ Supplier No. SA510192
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Rabbit Polyclonal Secondary Antibody
SA5-10192 targets Mouse IgG (H+L) in IDF and IP applications and shows reactivity with Mouse samples. The SA5-10192 immunogen is mouse IgG (heavy and light chains).
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Specifications
Antigen Mouse IgG (H+L)
Applications Immunodiffusion, Immunoprecipitation
Classification Polyclonal
Concentration 10.4 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.8
Host Species Rabbit
Immunogen mouse IgG (heavy and light chains).
Purification Method Ion-exchange chromatography
Quantity 20 mg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Mouse
Content And Storage -20°C
Product Type Secondary Antibody
Form Liquid
Isotype IgG