missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Mouse IgG (H+L) Secondary Antibody (Pre-adsorbed), Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-60654-0.5mg
This item is not returnable.
View return policy
Description
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Mouse samples. It is validated for Flow Cytometry, Immunomicroscopy.
Specifications
| IgG (H+L) | |
| Polyclonal | |
| Unconjugated | |
| 10 mM Sodium Phosphate, 0.15 M Sodium Chloride (pH 7.2), 1 % (w/v) BSA, Protease/IgG free with 0.05% Sodium Azide | |
| Purified Mouse IgG (H&L) | |
| 0.5 mg | |
| Secondary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Flow Cytometry, Immunomicroscopy | |
| 4.5mg/mL | |
| Flow Cytometry 1:20-1:2000, Immunomicroscopy 1:20-1:2000 | |
| Rabbit | |
| Affinity purified | |
| Adaptive Immunity, Immunology | |
| Based on IEP, this antibody reacts with heavy gamma chains on mouse IgG and light chains on all mouse immunoglobulins. This antibody has been pre-adsorbed against human serum proteins. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction