Learn More
Rabbit IgG Franction to Hamster IgG, Secondary Antibody, MP Biomedicals™
Shop All MP Biomedicals ProductsDescription
- Product is the lyophilized powder of rabbit IgG fraction to hamster IgG (whole molecule) and buffer salts.
- The IgG fraction is prepared from the specific rabbit antiserum by delipidation, ammonium sulfate fractionation and anion exchange chromatography
- This product is suitable for use as a precipitating antibody. It gives sharp precipitin lines in immunoelectrophoresis and immunodiffusion techniques.
- This product is suitable for conjugation with enzymes, radiolabels, fluorochromes or other markers, and for attachment to solid supports for use as an immunoadsorbant.
- This product is suitable for use as a primary reagent in enzyme immunoassays, fluorescein immunoassays and blot, cell or tissue immunostaining.
Specifications
Specifications
| Applications | Immunohistochemistry, ELISA |
| Conjugate | Unconjugated |
| Description | Product is the lyophilized powder of rabbit IgG fraction to hamster IgG (whole molecule) and buffer salts |
| Host Species | Rabbit |
| Purification Method | ammonium sulfate fractionation and anion exchange chromatography |
| Quantity | 2 mL |
| Regulatory Status | RUO |
| Primary or Secondary | Secondary |
| Reconstitution | Reconstitute product with 5.0mL of deionized or distilled water.gentle swirling may be used to speed rehydration. Avoid vigorous shaking of the reconstituted material. |
| Target Species | Hamster |
| Show More |