missing translation for 'onlineSavingsMsg'
Learn More

Rabeprazole EP Impurity H (Lansoprazole EP Impurity F ), Mikromol™

Product Code. 30461899
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30461899 4 x 25 mg -
30051067 25 mg 25mg
17904046 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30461899 Supplier Mikromol Supplier No. MM0474.0200254AMP
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Rabeprazole EP Impurity H (Lansoprazole EP Impurity F ), Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-[(RS)-[(4-Chloro-3-methylpyridin-2-yl)methyl]sulphinyl]-1H-benzimidazole
CAS 168167-42-8
Analyte or Component Names 2-[(RS)-[(4-Chloro-3-methylpyridin-2-yl)methyl]sulphinyl]-1H-benzimidazole
Shipping Condition Room Temperature
Molecular Formula C14 H12 Cl N3 O S
Certifications/Compliance ISO 17025
Synonym Lansoprazole Imp. F (EP), Rabeprazole Sodium Imp. H (EP), Rabeprazole Imp. H (EP), Rabeprazole USP Related Compound F, Rabeprazole USP RC F, 2-[(RS)-[(4-Chloro-3-methylpyridin-2-yl)methyl]sulphinyl]-1H-benzimidazole, Rabeprazole Related Compound F, Lansoprazole Impurity F, Rabeprazole Sodium Impurity H, Rabeprazole Impurity H
InChI Formula InChI=1S/C14H12ClN3OS/c1-9-10(15)6-7-16-13(9)8-20(19)14-17-11-4-2-3-5-12(11)18-14/h2-7H,8H2,1H3,(H,17,18)
Solution Type Neat
SMILES Cc1c(Cl)ccnc1CS(=O)c2nc3ccccc3[nH]2
IUPAC Name 2-[(4-chloro-3-methylpyridin-2-yl)methylsulfinyl]-1H-benzimidazole
Molecular Weight (g/mol) 305.78
Quantity 4 x 25 mg
Formula Weight 305.039 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.