missing translation for 'onlineSavingsMsg'
Learn More

rac 3-Hydroxydecanoic Acid Methyl Ester, TRC

Product Code. 30398355
Change view
Click to view available options
Quantity:
5 mg
25 mg
100 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30398355 25 mg 25mg
30393030 100 mg 100mg
30390174 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30398355 Supplier Toronto Research Chemicals Supplier No. TRCH93399525MG
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62675-82-5
Molecular Formula C11 H22 O3
Molecular Weight (g/mol) 202.29
Synonym (±)-Methyl 3-Hydroxydecanoate, Methyl 3-Hydroxydecanoate, Methyl β-Hydroxydecanoate
IUPAC Name methyl 3-hydroxydecanoate
SMILES CCCCCCCC(O)CC(=O)OC

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C11H22O3/c1-3-4-5-6-7-8-10(12)9-11(13)14-2/h10,12H,3-9H2,1-2H3
Formula Weight 202.1569
Chemical Name or Material Methyl 3-Hydroxydecanoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.