missing translation for 'onlineSavingsMsg'
Learn More

rac 4-(2,3-Dichlorophenyl)-3,4-dihydro-1(2H)-naphthalenone, TRC

Product Code. 30385235
Change view
Click to view available options
Quantity:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30385235 500 mg
30374738 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30385235 Supplier Toronto Research Chemicals Supplier No. TRCD435703500MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 152448-80-1
Molecular Formula C16H12Cl2O
Molecular Weight (g/mol) 291.17
Synonym 4-(2,3-Dichlorophenyl)-3,4-dihydro-1(2H)-naphthalenone
IUPAC Name 4-(2,3-dichlorophenyl)-3,4-dihydro-2H-naphthalen-1-one
SMILES Clc1cccc(C2CCC(=O)c3ccccc23)c1Cl

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C16H12Cl2O/c17-14-7-3-6-13(16(14)18)11-8-9-15(19)12-5-2-1-4-10(11)12/h1-7,11H,8-9H2
Formula Weight 290.0265
Chemical Name or Material rac 4-(2,3-Dichlorophenyl)-3,4-dihydro-1(2H)-naphthalenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.