missing translation for 'onlineSavingsMsg'
Learn More

rac-cis-3,4-Deschlorosertraline, TRC

Product Code. 30132878
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132878 100 mg 100mg
30010902 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30132878 Supplier Toronto Research Chemicals Supplier No. TRCD289840100MG
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52758-05-1
Molecular Formula C17 H19 N . Cl H
Molecular Weight (g/mol) 273.8
Synonym 1-Naphthalenamine, 1,2,3,4-tetrahydro-N-methyl-4-phenyl-, hydrochloride (1:1), (1R,4R)-rel-, 1-Naphthalenamine, 1,2,3,4-tetrahydro-N-methyl-4-phenyl-, hydrochloride, cis- (9CI), 1-Naphthalenamine, 1,2,3,4-tetrahydro-N-methyl-4-phenyl-, hydrochloride, cis-(±)-, (1RS,4RS)-N-Methyl-4-phenyl-1,2,3,4-tetrahydronaphthalen-1-amine Hydrochloride, Sertraline Hydrochloride Imp. B (EP) as Hydrochloride
IUPAC Name (1S,4S)-N-methyl-4-phenyl-1,2,3,4-tetrahydronaphthalen-1-amine;hydrochloride
SMILES Cl.CN[C@H]1CC[C@@H](c2ccccc2)c3ccccc13

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C17H19N.ClH/c1-18-17-12-11-14(13-7-3-2-4-8-13)15-9-5-6-10-16(15)17;/h2-10,14,17-18H,11-12H2,1H3;1H/t14-,17-;/m0./s1
Formula Weight 273.1284
Percent Purity >95%
Chemical Name or Material (1RS,4RS)-N-Methyl-4-phenyl-1,2,3,4-tetrahydronaphthalen-1-amine Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.