missing translation for 'onlineSavingsMsg'
Learn More

rac-cis-7-Hydroxy Pramipexole, TRC

Product Code. 30406395
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30406395 10 mg 10mg
30390532 25 mg 25mg
30394011 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30406395 Supplier Toronto Research Chemicals Supplier No. TRCH95214010MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (6RS,7RS)-2-Amino-6-(propylamino)-4,5,6,7-tetrahydrobenzothiazol-7-ol
CAS 1798014-87-5
Molecular Formula C10 H17 N3 O S
Quantity 10 mg
Synonym 7-Benzothiazolol, 2-amino-4,5,6,7-tetrahydro-6-(propylamino)-, (6R,7R)-rel-, rel-(6S,7S)-2-Amino-4,5,6,7-tetrahydro-6-(propylamino)-7-benzothiazolol, (6RS,7RS)-2-Amino-6-(propylamino)-4,5,6,7-tetrahydrobenzothiazol-7-ol, rac-cis-7-Hydroxypramipexole, (±)-
InChI Formula InChI=1S/C10H17N3OS/c1-2-5-12-6-3-4-7-9(8(6)14)15-10(11)13-7/h6,8,12,14H,2-5H2,1H3,(H2,11,13)/t6-,8-/m0/s1
SMILES CCCN[C@H]1CCc2nc(N)sc2[C@H]1O
IUPAC Name (6S,7S)-2-amino-6-(propylamino)-4,5,6,7-tetrahydro-1,3-benzothiazol-7-ol
Molecular Weight (g/mol) 227.327
Formula Weight 227.109
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.