missing translation for 'onlineSavingsMsg'
Learn More

rac-cis-N-Desmethyl Sertraline Hydrochloride, TRC

Product Code. 30008088
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30008088 50 mg 50mg
17992665 5 mg 5mg
17981965 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008088 Supplier Toronto Research Chemicals Supplier No. TRCD29210050MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91797-57-8
Molecular Formula C16 H15 Cl2 N . Cl H
Molecular Weight (g/mol) 328.66
Synonym (±)-cis-N-Desmethylsertraline Hydrochloride, cis-4-(3,4-Dichlorophenyl)-1,2,3,4-tetrahydro-1-naphthalenamine hydrochloride, (±)-4-(3,4-Dichlorophenyl)-1,2,3,4-tetrahydro-1-naphthalenamine hydrochloride, rac-Norsertraline Hydrochloride
IUPAC Name (1S,4S)-4-(3,4-dichlorophenyl)-1,2,3,4-tetrahydronaphthalen-1-amine;hydrochloride
SMILES Cl.N[C@H]1CC[C@@H](c2ccc(Cl)c(Cl)c2)c3ccccc13

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C16H15Cl2N.ClH/c17-14-7-5-10(9-15(14)18)11-6-8-16(19)13-4-2-1-3-12(11)13;/h1-5,7,9,11,16H,6,8,19H2;1H/t11-,16-;/m0./s1
Formula Weight 327.0348
Percent Purity >95%
Chemical Name or Material Norsertraline hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.