missing translation for 'onlineSavingsMsg'
Learn More

rac Epinephrine, TRC

Product Code. 18070220
Change view
Click to view available options
Quantity:
50 mg
1 g
Unit Size:
1g
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18070220 50 mg 50mg
18070830 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18070220 Supplier Toronto Research Chemicals Supplier No. TRCE58858550MG
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 329-65-7
Molecular Formula C9 H13 N O3
Molecular Weight (g/mol) 183.2
Synonym 1,2-Benzenediol, 4-[1-hydroxy-2-(methylamino)ethyl]-, 1,2-Benzenediol, 4-[1-hydroxy-2-(methylamino)ethyl]-, (±)-, Benzyl alcohol, 3,4-dihydroxy-α-[(methylamino)methyl]-, (±)- (8CI), (±)-Adrenaline, (±)-Epinephrine, 1-(3,4-Dihydroxy)phenyl-2-methylaminoethanol, 2-(Methylamino)-1-(3,4-dihydroxyphenyl)ethanol, DL-Adrenaline, Epirenamine, Racepinefrine, Racepinephrine, dl-Adrenaline, dl-Epinephrine
IUPAC Name 4-[1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol
SMILES CNCC(O)c1ccc(O)c(O)c1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C9H13NO3/c1-10-5-9(13)6-2-3-7(11)8(12)4-6/h2-4,9-13H,5H2,1H3
Formula Weight 183.0895
Chemical Name or Material DL-Epinephrine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.