missing translation for 'onlineSavingsMsg'
Learn More

rac Hydroxy Tizanidine Hydrochloride (Mixture of Tautomers), TRC

Product Code. 30003935
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003935 1 mg 1mg
30005439 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30003935 Supplier Toronto Research Chemicals Supplier No. TRCH9665001MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1794829-08-5
Molecular Formula C9 H8 Cl N5 O S . Cl H
Molecular Weight (g/mol) 306.17
Synonym 2-[(5-Chloro-2,1,3-benzothiadiazol-4-yl)amino]-4,5-dihydro-1H-imidazol-5-ol Hydrochloride
IUPAC Name 2-[(5-chloro-2,1,3-benzothiadiazol-4-yl)amino]-4,5-dihydro-1H-imidazol-5-ol;hydrochloride
SMILES Cl.OC1CN=C(Nc2c(Cl)ccc3nsnc23)N1

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C9H8ClN5OS.ClH/c10-4-1-2-5-8(15-17-14-5)7(4)13-9-11-3-6(16)12-9;/h1-2,6,16H,3H2,(H2,11,12,13);1H
Formula Weight 304.9905
Chemical Name or Material rac Hydroxy Tizanidine Hydrochloride (Mixture of Tautomers)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.