missing translation for 'onlineSavingsMsg'
Learn More

rac N-Acetyl Ephedrine, TRC

Product Code. 30138730
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138730 25 mg 25mg
30012142 5 mg 5mg
30010383 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138730 Supplier Toronto Research Chemicals Supplier No. TRCA17385025MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5661-42-7
Molecular Formula C12H17NO2
Molecular Weight (g/mol) 207.27
Synonym (R*,S*)-N-(2-Hydroxy-1-methyl-2-phenylethyl)-N-methylacetamide, erythro-N-(β-Hydroxy-α-methylphenethyl)-N-methylacetamide, N-Acetylephedrine;
IUPAC Name N-(1-hydroxy-1-phenylpropan-2-yl)-N-methylacetamide
SMILES OC([C@H](N(C)C(C)=O)C)C1=CC=CC=C1

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C12H17NO2/c1-9(13(3)10(2)14)12(15)11-7-5-4-6-8-11/h4-9,12,15H,1-3H3
Formula Weight 207.1259
Percent Purity >95%
Chemical Name or Material rac N-Acetyl Ephedrine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.