missing translation for 'onlineSavingsMsg'
Learn More

rac N-tert-Butoxycarbonyl Viloxazine, TRC

Product Code. 30332309
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30332309 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30332309 Supplier Toronto Research Chemicals Supplier No. TRCB690510100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material rac N-tert-Butoxycarbonyl Viloxazine
CAS 1330189-29-1
Molecular Formula C18H27NO5
Quantity 100 mg
Synonym (+/-)-2-N-tert-Butoxycarbonyl-(o-ethoxyphenoxymethyl)morpholine, N-tert-Butoxycarbonyl Catatrol, N-tert-Butoxycarbonyl Vicilan, N-tert-Butoxycarbonyl Viloxazin
InChI Formula InChI=1S/C18H27NO5/c1-5-21-15-8-6-7-9-16(15)23-13-14-12-19(10-11-22-14)17(20)24-18(2,3)4/h6-9,14H,5,10-13H2,1-4H3
SMILES CCOc1ccccc1OCC2CN(CCO2)C(=O)OC(C)(C)C
IUPAC Name tert-butyl 2-[(2-ethoxyphenoxy)methyl]morpholine-4-carboxylate
Molecular Weight (g/mol) 337.41
Formula Weight 337.1889
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.