missing translation for 'onlineSavingsMsg'
Learn More
Learn More
rac-Naproxen, LoGiCal
Specifications
Specifications
| Chemical Name or Material | rac Naproxen |
| CAS | 23981-80-8 |
| Analyte or Component Names | rac Naproxen |
| Shipping Condition | Room Temperature |
| Molecular Formula | C14 H14 O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | rac-Naproxen, 6-Methoxy-α±-methyl-2-naphthaleneacetic acid, (RS)-Naproxen, (±)-2-(6-Methoxy-2-naphthalenyl)propionic acid, (±)-2-(6-Methoxy-2-naphthyl)propionic acid, (±)-6-Methoxy-α±-methyl-2-naphthaleneacetic acid, (±)-Naproxen, 2-(6-Methoxy-2-naphthyl)propanoic acid, 2-(6-Methoxy-2-naphthyl)propionic acid, 2-(6-Methoxynaphthalen-2-yl)propanoic acid, 2-(6-Methoxynaphthalen-2-yl)propionic acid, 6-Methoxy-2-naphthyl-α±-methylacetic acid, DL-2-(6-Methoxy-2-naphthyl)propionic acid, DL-Naproxen, Racemic naproxen, α±-(6-Methoxy-2-naphthyl)propionic acid |
| InChI Formula | InChI=1S/C14H14O3/c1-9(14(15)16)10-3-4-12-8-13(17-2)6-5-11(12)7-10/h3-9H,1-2H3,(H,15,16) |
| Solution Type | Neat |
| SMILES | COc1ccc2cc(ccc2c1)C(C)C(=O)O |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?