missing translation for 'onlineSavingsMsg'
Learn More

rac-Nor Laudanosine Hydrochloride, TRC

Product Code. 30135875
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135875 1 g 1g
30138328 250 mg 250mg
30006236 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135875 Supplier Toronto Research Chemicals Supplier No. TRCN7100401G
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6429-04-5
Molecular Formula C20 H25 N O4 . Cl H
Molecular Weight (g/mol) 379.88
Synonym 1-[(3,4-Dimethoxyphenyl)methyl]-1,2,3,4-tetrahydro-6,7-dimethoxyisoquinoline Hydrochloride, 1,2,3,4-Tetrahydro-6,7-dimethoxy-1-veratrylisoquinoline Hydrochloride, (±)-N-Norlaudanosine Hydrochloride, (±)-Norlaudanosine Hydrochloride, (±)-Tetrahydropapaverine Hydrochloride, 1,2,3,4-Tetrahydropapaverine Hydrochloride, 1-(3,4-Dimethoxybenzyl)-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline Hydrochloride, Tetrahydropapaverine Hydrochloride;
IUPAC Name 1-[(3,4-dimethoxyphenyl)methyl]-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline;hydrochloride
SMILES Cl.COc1ccc(CC2NCCc3cc(OC)c(OC)cc23)cc1OC

Specifications

Quantity 1 g
InChI Formula InChI=1S/C20H25NO4.ClH/c1-22-17-6-5-13(10-18(17)23-2)9-16-15-12-20(25-4)19(24-3)11-14(15)7-8-21-16;/h5-6,10-12,16,21H,7-9H2,1-4H3;1H
Formula Weight 379.155
Chemical Name or Material rac-Nor Laudanosine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.