missing translation for 'onlineSavingsMsg'
Learn More

rac Norphenylephrine Hydrochloride (Phenylephrine Impurity A), TRC

Product Code. 30133275
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133275 100 mg 100mg
17973995 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133275 Supplier Toronto Research Chemicals Supplier No. TRCN825500100MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4779-94-6
Molecular Formula C8 H11 N O2 . Cl H
Molecular Weight (g/mol) 189.64
Synonym (1RS)-2-Amino-1-(3-hydroxyphenyl)ethanol Hydrochloride, Etilefrine Hydrochloride Imp. C (EP) as Hydrochloride, Benzenemethanol, α-(aminomethyl)-3-hydroxy-, hydrochloride (1:1), Benzenemethanol, α-(aminomethyl)-3-hydroxy-, hydrochloride (9CI), Benzyl alcohol, α-(aminomethyl)-m-hydroxy-, hydrochloride (6CI,8CI), (±)-Norphenylephrine hydrochloride, (±)-m-Octopamine hydrochloride, 1-(m-Hydroxyphenyl)-2-aminoethanol hydrochloride, 2-Amino-1-(3-hydroxyphenyl)ethanol hydrochloride, Coritat, DL-m-Octopamine hydrochloride, Depot-Novadral, Energona, Esbuphon, Molycor R, NSC 100733, NSC 297582, Norfenefrin, Norfenefrine hydrochloride, Norphenephrine hydrochloride, Norphenylephrine hydrochloride, Novadral, Stagural, Tonolift, Vingsal, WV 569, Zondel, dl-Norfenefrine hydrochloride, m-Octopamine hydrochloride, α-(Aminomethyl)-m-hydroxybenzyl alcohol hydrochloride
IUPAC Name 3-(2-amino-1-hydroxyethyl)phenol;hydrochloride
SMILES Cl.NCC(O)c1cccc(O)c1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C8H11NO2.ClH/c9-5-8(11)6-2-1-3-7(10)4-6;/h1-4,8,10-11H,5,9H2;1H
Formula Weight 189.0557
Percent Purity >95%
Chemical Name or Material Norfenefrine Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.