missing translation for 'onlineSavingsMsg'
Learn More

rac-Norsertraline Hydrochloride 0.1 mg/ml in Methanol (as free base), LoGiCal

Product Code. 30458695
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30458695 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30458695 Supplier LoGiCal Supplier No. LGCAMP0317.0202
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

rac-Norsertraline Hydrochloride 0.1 mg/ml in Methanol (as free base)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Norsertraline hydrochloride
CAS 91797-57-8
Analyte or Component Names Norsertraline hydrochloride
Shipping Condition Ice Pack (-20°C)
Molecular Formula C16 H15 Cl2 N . Cl H
Certifications/Compliance ISO 17034
Synonym (±)-cis-N-Desmethylsertraline Hydrochloride, cis-4-(3,4-Dichlorophenyl)-1,2,3,4-tetrahydro-1-naphthalenamine hydrochloride, (±)-4-(3,4-Dichlorophenyl)-1,2,3,4-tetrahydro-1-naphthalenamine hydrochloride, rac-Norsertraline Hydrochloride
InChI Formula InChI=1 S/C16H15Cl2N.ClH/c17-14-7-5-10(9-15(14)18)11-6-8-16(19)13-4-2-1-3-12(11)13;/h1-5,7,9,11,16 H,6,8,19H2;1 H/t11-,16-;/m0./s1
SMILES Cl.N[C@H]1 CC[C@@H](c2ccc(Cl)c(Cl)c2)c3ccccc13
IUPAC Name (1 S,4 S)-4-(3,4-dichlorophenyl)-1,2,3,4-tetrahydronaphthalen-1-amine;hydrochloride
Solvent or Matrix Methanol
Industry Type Forensics
Molecular Weight (g/mol) 328.66
Quantity 1 mL
Formula Weight 327.0348 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.