missing translation for 'onlineSavingsMsg'
Learn More

rac-Pinocembrin, TRC

Product Code. 30240861
Change view
Click to view available options
Quantity:
250 mg
2.5 g
5 g
Unit Size:
2.5g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30240861 2.5 g 2.5g
30240612 250 mg 250mg
30240709 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30240861 Supplier Toronto Research Chemicals Supplier No. TRCP4684952.5G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68745-38-0
Molecular Formula C15 H12 O4
Molecular Weight (g/mol) 256.25
Synonym Flavanone, 5,7-dihydroxy- (7CI), 2,3-Dihydro-5,7-dihydroxy-2-phenyl-4H-1-benzopyran-4-one, (±)-5,7-Dihydroxyflavanone, (±)-Pinocembrin, NSC 43318, rac-Pinocembrin
IUPAC Name 5,7-dihydroxy-2-phenyl-2,3-dihydrochromen-4-one
SMILES Oc1cc(O)c2C(=O)CC(Oc2c1)c3ccccc3

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C15H12O4/c16-10-6-11(17)15-12(18)8-13(19-14(15)7-10)9-4-2-1-3-5-9/h1-7,13,16-17H,8H2
Formula Weight 256.0736
Chemical Name or Material Pinocembrin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.