Learn More
rac Tenofovir-d6, TRC
Product Code. 30411710
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30411710 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30411710 Supplier Toronto Research Chemicals Supplier No. TRCT0185021MG
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Tenofovir-D6
CAS 1020719-94-1
Purity Grade Notes HPLC
Molecular Formula C9 2H6 H8 N5 O4 P
Quantity 1 mg
Synonym P-[[1-[(6-amino-9H-purin-9-yl)methyl-d2]ethoxy-1,2,2,2-d4]methyl]phosphonic acid
InChI Formula InChI=1S/C9H14N5O4P/c1-6(18-5-19(15,16)17)2-14-4-13-7-8(10)11-3-12-9(7)14/h3-4,6H,2,5H2,1H3,(H2,10,11,12)(H2,15,16,17)/i1D3,2D2,6D
SMILES [2H]C([2H])([2H])C([2H])(OCP(=O)(O)O)C([2H])([2H])n1cnc2c(N)ncnc12
IUPAC Name [1-(6-aminopurin-9-yl)-1,1,2,3,3,3-hexadeuteriopropan-2-yl]oxymethylphosphonic acid
Molecular Weight (g/mol) 293.25
Formula Weight 293.116
Percent Purity >95