missing translation for 'onlineSavingsMsg'
Learn More

rac-Trimethyl Ketoprofen (Mixture of (2’,3’4’) and (3’,4’,5’) Isomers), TRC

Product Code. 30456717
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30456717 25 mg 25mg
30457037 5 mg 5mg
30454456 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30456717 Supplier Toronto Research Chemicals Supplier No. TRCT79643025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material rac-Trimethyl Ketoprofen (Mixture of (2',3'4') and (3',4',5') Isomers)
Analyte or Component Names rac-Trimethyl Ketoprofen (Mixture of (2',3'4') and (3',4',5') Isomers)
Shipping Condition Room Temperature
Molecular Formula C19H20O3
InChI Formula InChI=1S/2C19H20O3/c1-11-8-17(9-12(2)13(11)3)18(20)16-7-5-6-15(10-16)14(4)19(21)22;1-11-8-9-17(13(3)12(11)2)18(20)16-7-5-6-15(10-16)14(4)19(21)22/h2*5-10,14H,1-4H3,(H,21,22)
Solution Type Neat
SMILES O=C(C1=CC=CC(C(C)C(O)=O)=C1)C2=C(C)C(C)=C(C)C=C2.O=C(C3=CC=CC(C(C)C(O)=O)=C3)C4=CC(C)=C(C)C(C)=C4
IUPAC Name 2-(3-(2,3,4-trimethylbenzoyl)phenyl)propanoic acid--2-(3-(3,4,5-trimethylbenzoyl)phenyl)propanoic acid (1/1)
Industry Type Pharmaceutical
Molecular Weight (g/mol) 296.36
Quantity 25 mg
Formula Weight 296.14
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.