missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rac1 Mouse anti-Human, Mouse, Rat, Clone: 4A4B11, Proteintech
Mouse Monoclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | Rac1 |
|---|---|
| Clone | 4A4B11 |
| Concentration | 0.7 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Immunofluorescence, Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16817715
|
Proteintech
66122-1-IG-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16807715
|
Proteintech
66122-1-IG-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The protein encoded by this gene is a GTPase which belongs to the RAS superfamily of small GTP-binding proteins. Members of this superfamily appear to regulate a diverse array of cellular events, including the control of cell growth, cytoskeletal reorganization, and the activation of protein kinases. Two transcript variants encoding different isoforms have been found for this gene.Specifications
| Rac1 | |
| 0.7 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| MIG5, p21 Rac1, RAC1, RAC1b, Ras like protein TC25, TC 25, TC25 | |
| Rac1 | |
| IgG2b | |
| Protein A | |
| Antibody |
| 4A4B11 | |
| Immunohistochemistry (Paraffin), Immunofluorescence, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| P63000, P63001, Q6RUV5 | |
| 19353, 363875, 5879 | |
| Rac1 Fusion Protein Ag17779 | |
| Primary | |
| -20°C | |
| RAC1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title