missing translation for 'onlineSavingsMsg'
Learn More

Racecadotril, TRC

Product Code. 30137736
Change view
Click to view available options
Quantity:
10 mg
100 mg
500 mg
Unit Size:
100mg
10mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137736 10 mg 10mg
30132835 500 mg 500mg
30008685 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137736 Supplier Toronto Research Chemicals Supplier No. TRCR07060010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81110-73-8
Molecular Formula C21 H23 N O4 S
Molecular Weight (g/mol) 385.48
Synonym Racecadotril, Benzyl [[(2RS)-2-[(acetylsulfanyl)methyl]-3-phenylpropanoyl]amino]acetate
IUPAC Name benzyl 2-[[2-(acetylsulfanylmethyl)-3-phenylpropanoyl]amino]acetate
SMILES CC(=O)SCC(Cc1ccccc1)C(=O)NCC(=O)OCc2ccccc2

Specifications

InChI Formula InChI=1S/C21H23NO4S/c1-16(23)27-15-19(12-17-8-4-2-5-9-17)21(25)22-13-20(24)26-14-18-10-6-3-7-11-18/h2-11,19H,12-15H2,1H3,(H,22,25)
Quantity 10 mg
Formula Weight 385.1348
Shipping Condition Room Temperature
Chemical Name or Material Racecadotril
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.