Learn More
Heidolph™ Rack For Reax Overhead Shaker
Product Code. 10490063 Shop All Heidolph Instruments Products
Change view
Click to view available options
Holds:
20 Test Tubes
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds Unit Size
10490063 20 Test Tubes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10490063 Supplier Heidolph™ Supplier No. 5492100000
Support a Safer, More Efficient Laboratory
Use promo code 28026
View all promotions
Estimated Shipment: 15-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Adaptor for 20 test tubes
Allows for simultaneously mixing 20 test tubes o.d. 10 to 18 mm with just one shaker
TRUSTED_SUSTAINABILITY
Specifications
Holds 20 Test Tubes
Product Type Rack
Diameter (Metric) 10 to 18 mm
No. of Positions 20