missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Raclopride
GREENER_CHOICE

Product Code. 30001288
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30001288 50 mg 50mg
18084802 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001288 Supplier Tocris Bioscience™ Supplier No. 1810/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent, selective D2/D3 antagonist

Selective and potent dopamine D2/D3 receptor antagonist (Ki values are 1.8, 3.5, 2400 and 18000 nM for D2, D3, D4 and D1 receptors respectively). Centrally active following systemic administration in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84225-95-6
Molecular Formula C15H20Cl2N2O3

Specifications

Quantity 50 mg
Formula Weight 347.24
Purity 98%
Molecular Weight (g/mol) 347.236
Chemical Name or Material Raclopride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.