missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ractopamine-d9 HCl (2-hydroxyethyl-1,1,2-d3; 1-methyl-d3-propyl-1,2,2-d3) (mixture of diastereomers), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Ractopamine-D9 (2-hydroxyethyl-1,1,2-D3,1-(methyl-D3)propyl-1,2,2-D3) Hydrochloride |
| Molecular Formula | C18 D9 H14 N O3 . Cl H |
| Quantity | 0.005 g |
| Synonym | 4-Hydroxy-α-[[[3-(4-hydroxyphenyl)-1-(methyl-d3)propyl-1,2,2-d3]amino]methyl-d2]benzenemethan-d-ol hydrochloride, Ractopamine-d9 hydrochloride, Ractopamine-D9 (2-hydroxyethyl-1,1,2-D3,1-(methyl-D3)propyl-1,2,2-D3) hydrochloride, 4-[1,2,2-Trideuterio-2-[[1,1,1,2,3,3-Hexadeuterio-4-(4-hydroxyphenyl)butan-2-yl]amino]-1-hydroxyethyl]phenol hydrochloride |
| InChI Formula | InChI=1 S/C18H23NO3.ClH/c1-13(2-3-14-4-8-16(20)9-5-14)19-12-18(22)15-6-10-17(21)11-7-15;/h4-11,13,18-22 H,2-3,12H2,1H3;1 H/i1D3,2D2,12D2,13 D,18 D; |
| SMILES | Cl.[2 H]C([2 H])([2 H])C([2 H])(NC([2 H])([2 H])C([2 H])(O)c1ccc(O)cc1)C([2 H])([2 H])Cc2ccc(O)cc2 |
| IUPAC Name | 4-[2,2,3,4,4,4-hexadeuterio-3-[[1,1,2-trideuterio-2-hydroxy-2-(4-hydroxyphenyl)ethyl]amino]butyl]phenol;hydrochloride |
| Molecular Weight (g/mol) | 346.897 |
| Formula Weight | 346.201 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto