Learn More
RAD23B Monoclonal Antibody (OTI7F11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA804862S
Description
RAD23B Monoclonal antibody specifically detects RAD23B in Human samples. It is validated for Western BlotSpecifications
| RAD23B | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q15013 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 9587 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI7F11 | |
| Unconjugated | |
| MAD2L1BP | |
| 0610007D13Rik; AV001138; HHR23B; HR23B; Mhr23b; P58; RAD23 homolog B (S. cerevisiae); RAD23 homolog B, nucleotide excision repair protein; RAD23, yeast homolog of, B; RAD23B; RAD23b homolog; RAD23b homolog (S. cerevisiae); UV excision repair protein RAD23 homolog B; XP-C repair complementing complex 58 kDa; XP-C repair complementing protein; XP-C repair-complementing complex 58 kDa protein | |
| Full length human recombinant protein of human MAD2L1BP produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.