Learn More
Invitrogen™ RAD51D Recombinant Rabbit Monoclonal Antibody (24GB5175)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557203
 View more versions of this product
Product Code. 30274806
  • 500.00€ / 100µL
Description
RAD51D Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Five human RAD 51 homologues have been identified: XRCC2, XRCC3, RAD51B, RAD51C, and RAD51D. Each of these homologues interacts with one or more of the others, with all of the proteins involved in one complex or multiple smaller complexes. RAD51D interacts with XRCC2 and RAD51C. RAD51D's expression pattern and sequence similarity to other RAD51 family members most likely make it part of a complex of proteins required for DNA repair and meiotic recombination.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RAD51D
Recombinant Monoclonal
1.4 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O55230, O75771
RAD51D
A synthesized peptide derived from human Rad51D (aa 1-120).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
24GB5175
Unconjugated
RAD51D
BROVCA4; DNA repair protein RAD51 homolog 4; DNA repair protein RAD51-like 4; HsTRAD; R51H3; RAD51 homolog D; RAD51 paralog D; Rad51d; RAD51L3; RAD51-like 3; RAD51-like protein 3; recombination repair protein; TRAD
Rabbit
Affinity chromatography
RUO
19364, 303375, 5892
-20°C