Learn More
RAD52 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 28045-1-AP-20UL
 View more versions of this product
Product Code. 16825975
  • 145.00€ / 20µL
Description
DNA double-strand breaks can be generated by exposure to ionizing radiation or through endogenous cellular reactions. Double-strand breaks are generally thought to be repaired via one of two distinct mechanisms: homologous recombination (HR) or non-homologous end joining (NHEJ). Homologous recombination requires the activity of the members of the RAD52 epistasis group, which includeRAD50, RAD51, RAD52, RAD54, RDH54/TID1, RAD55, RAD57, RAD59, MRE11, and XRS2. RAD52 can catalyze strand annealing reactions, and can form oligomers that attach to ends of single-stranded DNA.
Specifications
Specifications
RAD52
Polyclonal
Unconjugated
RAD52
RAD52, RAD52 homolog (S. cerevisiae)
Rabbit
Antigen Affinity Chromatography
RUO
5893
-20°C
Liquid
Immunoprecipitation, Western Blot
0.4 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P43351
RAD52
RAD52 Fusion Protein Ag27619