missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Radixin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13790-1-AP-20UL
This item is not returnable.
View return policy
Description
Ezrin, radixin and moesin (ERM) proteins are widely distributed proteins located in the cellular cortex, in microvilli and adherens junctions. They are mediate linkage of actin cytoskeleton to plasma membrane in many cells. ERM proteins contain an N-terminal FERM (4.1/ezrin/radixin/moesin) domain that binds to phosphatidylinositol-(4, 5)phosphate and cellular membrane proteins. The ERM, particularly ezrin, is important for reconstructing cell-surface architecture during T cell activation. Despite the high degree of homology, the three proteins exhibit a distinct receptor-specific pattern of phosphorylation. ERM activity is regulated in part by phosphorylation at a C-terminal threonine (ezrin-Thr567, radixin-Thr564, moesin-Thr558).Specifications
| Radixin | |
| Polyclonal | |
| Unconjugated | |
| RDX | |
| DFNB24, radixin, RDX | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 19684, 315655, 5962 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin), Flow Cytometry, Immunofluorescence, Immunocytochemistry | |
| 0.31 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P26043, P35241 | |
| Rdx | |
| Radixin Fusion Protein Ag4813 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction