missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RALB Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12340-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a GTP-binding protein that belongs to the small GTPase superfamily and Ras family of proteins. GTP-binding proteins mediate the transmembrane signaling initiated by the occupancy of certain cell surface receptors.Specifications
| RALB | |
| Polyclonal | |
| Unconjugated | |
| RALB | |
| RALB, Ras related protein Ral B | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 5899, 64143 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P11234, Q9JIW9 | |
| RALB | |
| RALB Fusion Protein Ag2992 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction