missing translation for 'onlineSavingsMsg'
Learn More

Ramelteon Metabolite M-II (mixture of R and S at the hydroxy position), TRC

Product Code. 30409888
Change view
Click to view available options
Quantity:
1 mg
5 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30409888 1 mg
30408841 5 mg
30423548 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30409888 Supplier Toronto Research Chemicals Supplier No. TRCR1100601MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 896736-21-3
Molecular Formula C16 H21 N O3
Molecular Weight (g/mol) 275.34
Synonym Ramelteon Metabolite M-Ii
IUPAC Name 2-hydroxy-N-[2-(2,6,7,8-tetrahydro-1H-cyclopenta[e][1]benzofuran-8-yl)ethyl]propanamide
SMILES CC(O)C(=O)NCCC1CCc2ccc3OCCc3c12

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C16H21NO3/c1-10(18)16(19)17-8-6-12-3-2-11-4-5-14-13(15(11)12)7-9-20-14/h4-5,10,12,18H,2-3,6-9H2,1H3,(H,17,19)
Formula Weight 275.1521
Percent Purity >95%
Chemical Name or Material 2-hydroxy-N-[2-(2,6,7,8-tetrahydro-1H-cyclopenta[e][1]benzofuran-8-yl)ethyl]propanamide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.