missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RanBP16 Recombinant Rabbit Monoclonal Antibody (HL2510)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556853
This item is not returnable.
View return policy
Description
RanBP16 Recombinant Monoclonal Antibody for Western Blot, ICC/IF
The transport of protein and large RNAs through the nuclear pore complexes is an energy-dependent and regulated process. The import of proteins with a nuclear localization signal is accomplished by recognition of one or more clusters of basic amino acids by the importin-alpha/beta complex; see MIM 600685 and MIM 602738. The small GTPase RAN plays a key role in NLS-dependent protein import. RAN-binding protein-16 is a member of the importin-beta superfamily of nuclear transport receptors.
Specifications
| RanBP16 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q9EPK7, Q9UIA9 | |
| Xpo7 | |
| Recombinant fragment of Human RanBP16. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| HL2510 | |
| Unconjugated | |
| Xpo7 | |
| 4930506C02Rik; BB164534; EXP7; exportin 7; exportin-7; Kiaa0745; mKIAA0745; RAN binding protein 16; ran-binding protein 16; Ranbp16; Xpo7 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 23039, 65246 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction