Learn More
Raney™-Nickel, Activated Catalyst, 50% slurry in water
Product Code. 10172953 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10172953 500 g 500g
10243353 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10172953 Supplier Thermo Scientific Acros Supplier No. 395925000
  • ONLINE EXCLUSIVE PRICE
    234.00€
    210.65€ / 500g
    Save 23.35€
    9% Off
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
missing translation for 'validMainframeMsg'
CAS: 7440-02-0 | Ni | 58.69 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7440-02-0 , 7732-18-5 , 7429-90-5
Molecular Formula Ni
Molecular Weight (g/mol) 58.69
MDL Number MFCD00011137 MFCD06798735
InChI Key PXHVJJICTQNCMI-UHFFFAOYSA-N
Synonym raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
PubChem CID 935
ChEBI CHEBI:28112
IUPAC Name nickel
SMILES [Ni]
Specifications
Quantity 500 g
Packaging Plastic bottle
Form Slurry
Formula Weight 58.69
Chemical Name or Material Raney™-Nickel, Activated Catalyst