missing translation for 'onlineSavingsMsg'
Learn More

RARRES1 Monoclonal Antibody (OTI1C9), TrueMAB™, OriGene

Mouse Monoclonal Antibody

Brand:  Origene Technologies TA506082S

Encompass_Preferred

 View more versions of this product

Product Code. 17555382

  • 242.00€ / 30µL
Only null left
Estimated Shipment: 12-01-2026
Add to basket
This item is not returnable. View return policy

Description

Description

RARRES1 Monoclonal antibody specifically detects RARRES1 in Human samples. It is validated for Western Blot
Specifications

Specifications

RARRES1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q02750
Mouse
Affinity chromatography
RUO
5604
-20° C, Avoid Freeze/Thaw Cycles
IgG2b
Western Blot
OTI1C9
Unconjugated
Map2k1
5430417P09Rik; AI662122; latexin-like; LXNL; PEIG1; PERG-1; phorbol ester-induced gene 1 protein; Rarres1; RAR-responsive protein TIG1; retinoic acid receptor responder (tazarotene induced) 1; retinoic acid receptor responder 1; retinoic acid receptor responder protein 1; Tazarotene-induced gene 1 protein; tazarotene-induced protein 1; TIG1
Full length human recombinant protein of human MAP2K1 produced in HEK293T cell.
30 μL
Primary
Human
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.