missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RARS2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA576405
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
The protein encoded by this gene is an arginyl-tRNA synthetase that is found in the mitochondrial matrix. Defects in this gene are a cause of pontocerebellar hypoplasia type 6 (PCH6).
Specifications
| RARS2 | |
| Polyclonal | |
| Unconjugated | |
| RARS2 | |
| 1500002I10Rik; arginine--tRNA ligase; Arginyl-tRNA synthetase; arginyl-tRNA synthetase 2, mitochondrial; arginyl-tRNA synthetase 2, mitochondrial (putative); ArgRS; DALRD2; dJ382I10.6; PCH6; PRO1992; probable arginine--tRNA ligase, mitochondrial; probable arginyl-tRNA synthetase, mitochondrial; Rars2; RARSL | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 109093, 57038 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q3U186, Q5T160 | |
| RARS2 | |
| Recombinant fusion protein of human RARS2. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction