missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat Serpin A12 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF5347-SP
This item is not returnable.
View return policy
Description
Serpin A12 Polyclonal specifically detects Serpin A12 in Rat samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Serpin A12 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| antitrypsin), member 12, member 12, OL-64, OL-64Visceral adipose-specific serpin, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, serpin A12, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), Vaspin, Visceral adipose tissue-derived serine protease inhibitor | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 145264 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunoprecipitation | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Q8R4Z1 | |
| Serpina12 | |
| Mouse myeloma cell line NS0-derived recombinant rat Serpin A12 Leu20-Pro411 Accession # Q8R4Z1 | |
| 25 μg | |
| Primary | |
| Detects rat Serpin A12 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 5% cross-reactivity with recombinant human Serpin A12 is observed. | |
| Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction