missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat BOK Antibody, R&D Systems™

Rabbit Polyclonal Antibody
Brand: R&D Systems AF825
This item is not returnable.
View return policy
Description
BOK Polyclonal specifically detects BOK in Rat samples. It is validated for Western Blot.
Specifications
| BOK | |
| Polyclonal | |
| Unconjugated | |
| Bcl2-L-9, BCL2L9bcl-2-related ovarian killer protein, Bcl-2-like protein 9, BCL2-related ovarian killer, BOKL, hBOK, MGC4631 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 666 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.8 ug/mL | |
| BOK | |
| KLH-coupled rat BOK synthetic peptide ARLLRAGLSWSAPERASPAPGC | |
| 80 μg | |
| Primary | |
| Detects rat BOK in Western blots. | |
| Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction